Invitrogen
Manufacturer Code:PA521468
Catalog # PIPA521468
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 222 and 469 of CSE1L (Uniprot IDNo.P55060) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAS CSE1 XPO2 cellular apoptosis susceptibility protein chromosome segregation 1-like protein exp2 exportin-2 importin-alpha re-exporter; Cellular apoptosis susceptibility protein; Chromosome segregation 1-like protein; CSE1 chromosome segregation 1 like; CSE1 chromosome segregation 1-like; Exp2; Exportin-2; Importin-alpha re-exporter
Gene Aliases: 2610100P18Rik; AA407533; Capts; CAS; CSE1; CSE1L; XPO2
UniProt ID: (Human) P55060, (Mouse) Q9ERK4
Entrez Gene ID: (Human) 1434, (Mouse) 110750, (Rat) 362273
Molecular Function: G-protein enzyme modulator small GTPase transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.