Proteintech
Manufacturer Code:132121AP150UL
Catalog # 50-172-8576
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CARTPT Fusion Protein Ag4004 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CART CART prepropeptide CARTPT; CART(1-39); CART(1-52); CART(42-89); CART(55-102); CART(62-102); cocaine and amphetamine regulated transcript; Cocaine- and amphetamine-regulated transcript protein
Gene Aliases: CART; CARTPT
UniProt ID: (Human) Q16568, (Mouse) P56388, (Rat) P49192
Entrez Gene ID: (Human) 9607, (Mouse) 27220, (Rat) 29131
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.