Novus Biologicals
Manufacturer Code:NBP212069AF700
Catalog # NB212069700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0124-7C11 |
Immunogen | Human CARTPT |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: CART prepropeptide CARTcocaine and amphetamine regulated transcript cocaine- and amphetamine-regulated transcript protein; CART(1-39); CART(42-89); Cocaine- and amphetamine-regulated transcript protein
Gene Aliases: CART; CARTPT
UniProt ID: (Human) Q16568
Entrez Gene ID: (Human) 9607
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.