Novus Biologicals
Manufacturer Code:NBP271771G
Catalog # NBP271771G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3D7 |
Immunogen | Full length recombinant protein of human CARKL (NP_037408) produced in HEK293T cells. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: carbohydrate kinase-like Carbohydrate kinase-like protein CARKLEC 2.7.1.14 sedoheptulokinase SHKFLJ32478; Carbohydrate kinase-like protein; Sedoheptulokinase; SHK
Gene Aliases: CARKL; SHK; SHPK
UniProt ID: (Human) Q9UHJ6
Entrez Gene ID: (Human) 23729
Molecular Function: carbohydrate kinase kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.