Invitrogen
Manufacturer Code:PA5101269
Catalog # PIPA5101269
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CARKL corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4930431K22Rik AI194947 AW260459 Carbohydrate kinase-like protein Carkl sedoheptulokinase SHK SHPK; Carbohydrate kinase-like protein; Sedoheptulokinase; SHK
Gene Aliases: 4930431K22Rik; AI194947; AW260459; CARKL; SHK; SHPK
UniProt ID: (Human) Q9UHJ6, (Mouse) Q9D5J6
Entrez Gene ID: (Human) 23729, (Rat) 287479, (Mouse) 74637
Molecular Function: carbohydrate kinase kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.