Invitrogen
Manufacturer Code:PA529189
Catalog # PIPA529189
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 36 and 249 of Human CARD9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CANDF2 hCARD9 caspase recruitment domain-containing protein 9; caspase recruitment domain family, member 9; Caspase recruitment domain-containing protein 9; hCARD9
Gene Aliases: CANDF2; CARD9; Gm782; hCARD9
UniProt ID: (Human) Q9H257, (Mouse) A2AIV8
Entrez Gene ID: (Human) 64170, (Mouse) 332579
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.