Proteintech
Manufacturer Code:106691AP150UL
Catalog # 50-172-6497
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CARD9 Fusion Protein Ag1057 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CARD9 hCARD9; caspase recruitment domain family, member 9; caspase recruitment domain protein 9; Caspase recruitment domain-containing protein 9; hCARD9; rCARD9
Gene Aliases: CANDF2; CARD9; Gm782; hCARD9
UniProt ID: (Human) Q9H257, (Rat) Q9EPY0, (Mouse) A2AIV8
Entrez Gene ID: (Human) 64170, (Rat) 64171, (Mouse) 332579
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.