Invitrogen
Manufacturer Code:PA559852
Catalog # PIPA559852
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CARD8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apoptotic protein NDPP1; CARD inhibitor of NF-kappaB-activating ligands; CARD-inhibitor of NF-kappa-B-activating ligand; CARDINAL; CARDINAL DACAR DAKAR NDPP NDPP1 TUCAN CARD inhibitor of NF-kappaB-activating ligands apoptotic protein NDPP1 caspase recruitment domain-containing protein 8 tumor up-regulated CARD-containing antagonist of CASP9 tumor up-regulated CARD-containing antagonist of caspase nine; caspase recruitment domain family, member 8; Caspase recruitment domain-containing protein 8; DACAR; TUCAN; Tumor up-regulated CARD-containing antagonist of CASP9; tumor up-regulated CARD-containing antagonist of caspase nine
Gene Aliases: CARD8; CARDINAL; DACAR; DAKAR; KIAA0955; NDPP; NDPP1; TUCAN
UniProt ID: (Human) Q9Y2G2
Entrez Gene ID: (Human) 22900
Molecular Function: cysteine protease enzyme modulator hydrolase protease protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.