Invitrogen
Manufacturer Code:MA535829
Catalog # PIMA535829
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1680 |
Immunogen | A synthesized peptide derived from human CARD11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 0610008L17Rik 2410011D02Rik BENTA BIMP3 Card11 CARD-containing MAGUK protein 1 Carma 1 CARMA1 Caspase recruitment domain-containing protein 11 IMD11 MGC133069 PPBL; bcl10-interacting maguk protein 3; CARD-containing MAGUK protein 1; Carma 1; caspase recruitment domain family, member 11; Caspase recruitment domain-containing protein 11
Gene Aliases: 0610008L17Rik; 2410011D02Rik; BENTA; BIMP3; CARD11; CARMA1; IMD11; PPBL
UniProt ID: (Human) Q9BXL7, (Mouse) Q8CIS0
Entrez Gene ID: (Human) 84433, (Mouse) 108723
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.