Invitrogen
Manufacturer Code:PA520032
Catalog # PIPA520032
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids at the C-terminus of human CARMA1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bcl10-interacting maguk protein 3; BIMP3 CARMA1 CARD-containing MAGUK protein 1 bcl10-interacting maguk protein 3 card-maguk protein 1 carma 1 caspase recruitment domain-containing protein 11; CARD-containing MAGUK protein 1; Carma 1; caspase recruitment domain family, member 11; Caspase recruitment domain-containing protein 11
Gene Aliases: 0610008L17Rik; 2410011D02Rik; BENTA; BIMP3; CARD11; CARMA1; IMD11; PPBL
UniProt ID: (Human) Q9BXL7, (Mouse) Q8CIS0
Entrez Gene ID: (Human) 84433, (Mouse) 108723
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.