Invitrogen
Manufacturer Code:PA518145
Catalog # PIPA518145
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (QRKTIWVDEDQL) corresponding to the C-terminus amino acids of CARD11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: bcl10-interacting maguk protein 3; CARD-containing MAGUK protein 1; CARD11 caspase recruitment domain family member 11 BIMP3 CARMA1 MGC133069 card-maguk protein 1 bcl10-interacting maguk protein 3; Carma 1; caspase recruitment domain family, member 11; Caspase recruitment domain-containing protein 11
Gene Aliases: BENTA; BIMP3; CARD11; CARMA1; IMD11; PPBL
UniProt ID: (Human) Q9BXL7
Entrez Gene ID: (Human) 84433
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.