Abnova
Manufacturer Code:PAB12907
Catalog # 89-116-179
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of human CARD11. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of CARD11.
For Research Use Only
Protein Aliases: bcl10-interacting maguk protein 3; BIMP3; CARD-containing MAGUK protein 1; Carma 1; CARMA1; caspase recruitment domain family, member 11; Caspase recruitment domain-containing protein 11; MGC133069
Gene Aliases: BENTA; BIMP3; CARD11; CARMA1; IMD11; PPBL
UniProt ID: (Human) Q9BXL7
Entrez Gene ID: (Human) 84433
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.