Invitrogen
Manufacturer Code:PA533167
Catalog # PIPA533167
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunodiffusion (IDF) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Purified human carbonic anhydrase II (CAII) prepared from erythrocytes. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4°C or -20°C if preferred |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: CA II; CA-II; CA-II CAII Car2CAC carbonate dehydratase II carbonic anhydrase 2 carbonic anhydrase B carbonic anhydrase C carbonic dehydratase; CAC; Carbonate dehydratase II; Carbonic anhydrase 2; carbonic anhydrase B; Carbonic anhydrase C; Carbonic anhydrase II; carbonic dehydratase; epididymis luminal protein 76; epididymis secretory protein Li 282
Gene Aliases: AI131712; CA-II; CA2; CAC; CAII; Car-2; Car2; HEL-76; HEL-S-282; Ltw-5; Lvtw-5
UniProt ID: (Human) P00918, (Mouse) P00920
Entrez Gene ID: (Human) 760, (Mouse) 12349
Molecular Function: dehydratase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.