R&D Systems
Manufacturer Code:MAB3724SP
Catalog # MAB3724SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 331930 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse CAR/NR1I3 Leu20-Gly237 Accession # P97792 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CAR; CARCAR1 Constitutive activator of retinoid response constitutive active receptor Constitutive active response Constitutive androstane receptor MB67 MGC150433 MGC97144 MGC97209 NR1I3 nuclear receptor subfamily 1 group I member 3 nuclear receptor subfamily 1 group I member 3 orphan nuclear hormone receptor Orphan nuclear receptor MB67; Constitutive activator of retinoid response; constitutive active receptor; Constitutive active response; constitutive androstane nuclear receptor variant 2; constitutive androstane nuclear receptor variant 3; constitutive androstane nuclear receptor variant 4; constitutive androstane nuclear receptor variant 5; Constitutive androstane receptor; Nuclear receptor subfamily 1 group I member 3; nuclear receptor subfamily 1, group I, member 3; orphan nuclear hormone receptor; Orphan nuclear receptor MB67
Gene Aliases: CAR; CAR1; MB67; NR1I3
UniProt ID: (Human) Q14994
Entrez Gene ID: (Human) 9970
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.