Invitrogen
Manufacturer Code:PA547364
Catalog # PIPA547364
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse CXADR Leu20-Gly237 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CAR; CAR CAR4/6 HCAR 46 kD coxsackievirus and adenovirus receptor (CAR) protein CVB3-binding protein HCVADR coxsackievirus B-adenovirus receptor coxsackievirus and adenovirus receptor; Coxsackievirus and adenovirus receptor homolog
Gene Aliases: 2610206D03Rik; AU016810; AW553441; CAR; Cxadr; MCAR; MCVADR
UniProt ID: (Mouse) P97792
Entrez Gene ID: (Mouse) 13052
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.