Proteintech
Manufacturer Code:159481AP150UL
Catalog # 50-173-0591
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CAPZA2 Fusion Protein Ag8544 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAPPA2 CAPZ CapZ alpha 2 CAPZA2; capping protein (actin filament) muscle Z-line, alpha 2; capping protein alpha 2; CapZ alpha-2; F-actin capping protein alpha-2 subunit; F-actin-capping protein subunit alpha-2
Gene Aliases: 1110053K06Rik; AW208808; CAPPA2; CAPZ; CAPZA2
UniProt ID: (Human) P47755, (Rat) Q3T1K5, (Mouse) P47754
Entrez Gene ID: (Human) 830, (Rat) 493810, (Mouse) 12343
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.