Proteintech
Manufacturer Code:207661AP150UL
Catalog # 50-173-3069
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CAPRIN2 Fusion Protein Ag14745 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1q domain containing 1; C1q domain-containing protein 1; C1QDC1 caprin 2 caprin family member 2 CAPRIN2 EEG 1 EEG1 KIAA1873 Protein EEG 1 RNA granule protein 140 RNG140; caprin family member 2; Caprin-2; caprin2; Cytoplasmic activation/proliferation-associated protein 2; Gastric cancer multidrug resistance-associated protein; Protein EEG-1; RNA granule protein 140
Gene Aliases: C1QDC1; CAPRIN2; EEG-1; EEG1; KIAA1873; RNG140
UniProt ID: (Human) Q6IMN6, (Mouse) Q05A80
Entrez Gene ID: (Human) 65981, (Rat) 686779, (Mouse) 232560
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.