Abnova
Manufacturer Code:H00010753A01
Catalog # 89-106-398
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 591 a.a. to 690 a.a) partial recombinant protein with GST tag.,CAPN9 (NP_006606 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant CAPN9.
For Research Use Only
Protein Aliases: Calpain-9; Digestive tract-specific calpain; GC36; nCL-4; New calpain 4; novel calpain large subunit-4; Protein CG36
Gene Aliases: CAPN9; GC36; nCL-4; NCL4
UniProt ID: (Human) O14815
Entrez Gene ID: (Human) 10753
Molecular Function: annexin calcium-binding protein calmodulin cysteine protease hydrolase intracellular calcium-sensing protein protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.