Abnova
Manufacturer Code:H00023473K
Catalog # 89-201-373
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human CAPN7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human CAPN7 peptide using ARM Technology.
For Research Use Only
Protein Aliases: calpain like protease; Calpain-7; CALPAIN7 FLJ36423 PALBH; homolog of Aspergillus Nidulans PALB; PalB homolog; PalBH
Gene Aliases: CALPAIN7; CAPN7; PALBH
UniProt ID: (Human) Q9Y6W3
Entrez Gene ID: (Human) 23473
Molecular Function: annexin calcium-binding protein calmodulin cysteine protease hydrolase intracellular calcium-sensing protein protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.