Abnova
Manufacturer Code:H00000726M02A
Catalog # 89-016-383
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3C10 |
Immunogen | CAPN5 (NP_004046 523 a.a. ~ 622 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant CAPN5.
For Research Use Only
Protein Aliases: Calpain htra-3; Calpain-5; FLJ46245; HTRA3; nCL-3; New calpain 3; testis tissue sperm-binding protein Li 91mP
Gene Aliases: ADNIV; CAPN5; HTRA3; nCL-3; NCL3; VRNI
UniProt ID: (Human) O15484
Entrez Gene ID: (Human) 726
Molecular Function: annexin calcium-binding protein calmodulin cysteine protease hydrolase intracellular calcium-sensing protein protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.