Proteintech
Manufacturer Code:121641AP150UL
Catalog # 50-172-7761
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CANT1 Fusion Protein Ag2805 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: apyrase 1, homolog (C. lectularius); Apyrase homolog; Apyrase homolog CANT1 SCAN 1 SHAPY; Ca2+-dependent endoplasmic reticulum nucleoside diphosphatase; ectonucleoside triphosphate diphosphohydrolase 8; micromelic dwarfism with vertebral and metaphyseal abnormalities and advanced carpotarsal ossification; Putative MAPK-activating protein PM09; Putative NF-kappa-B-activating protein 107; SCAN-1; soluble Ca-activated nucleotidase, isozyme 1; Soluble calcium-activated nucleotidase 1; soluble calcium-activated nucleotidase SCAN-1
Gene Aliases: 5830420C20Rik; Apy1h; CANT1; D11Bwg0554e; DBQD; DBQD1; Entpd8; SCAN-1; SCAN1; SHAPY; Srapy
UniProt ID: (Human) Q8WVQ1, (Mouse) Q8VCF1, (Rat) Q8K4Y7
Entrez Gene ID: (Human) 124583, (Mouse) 76025, (Rat) 246272
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.