Novus Biologicals
Manufacturer Code:NBP176864DL594
Catalog # N176864D594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of the human Cathelicidin. The immunogen is located within amino acids 50 - 100 of Cathelicidin. |
Conjugate | DyLight 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
Protein Aliases: 18 kDa cationic antimicrobial protein; 18 kDa cationic antimicrobial protein CAP18CRAMP cathelicidin antimicrobial peptide FALL-39 FALL39CAP-18 hCAP-18 LL37; Antibacterial peptide FALL-39; Antibacterial peptide LL-37; CAP-18; Cathelicidin antimicrobial peptide; FALL-39 peptide antibiotic
Gene Aliases: CAMP; CAP-18; CAP18; CRAMP; FALL-39; FALL39; HSD26; LL37
UniProt ID: (Human) P49913
Entrez Gene ID: (Human) 820
Molecular Function: antibacterial response protein defense/immunity protein enzyme modulator protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.