Novus Biologicals
Manufacturer Code:NBP270331PECY55
Catalog # N270331PEC5
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A3 |
Immunogen | Full length recombinant protein of human CAMLG (NP_001736) produced in HEK293T cells. |
Conjugate | PE-Cyanine5.5 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: calcium modulating ligand calcium signal-modulating cyclophilin ligand calcium-signal modulating cyclophilin ligand CAMLcalcium-modulating cyclophilin ligand cyclophilin B-binding protein MGC163197; Calcium signal-modulating cyclophilin ligand; calcium-modulating cyclophilin ligand; calcium-signal modulating cyclophilin ligand; CAML; cyclophilin B-binding protein
Gene Aliases: CAML; CAMLG; GET2
UniProt ID: (Human) A1L3Y3
Entrez Gene ID: (Human) 819
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.