Abnova
Manufacturer Code:H00000817M02
Catalog # 89-016-419
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1A8 |
Immunogen | CAMK2D (AAH32784 301 a.a. ~ 410 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant CAMK2D.
For Research Use Only
Protein Aliases: calcium/calmodulin-dependent protein kinase (CaM kinase) II delta; calcium/calmodulin-dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase type II delta chain; Calcium/calmodulin-dependent protein kinase type II subunit delta; CaM kinase II delta subunit; CaM kinase II subunit delta; CaM-kinase II delta chain; CaMK-II delta subunit; caMK-II subunit delta; CAMKD; DKFZp686G23119; DKFZp686I2288; MGC44911
Gene Aliases: CAMK2D; CAMKD
UniProt ID: (Human) Q13557
Entrez Gene ID: (Human) 817
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.