Invitrogen
Manufacturer Code:MA525045
Catalog # PIMA525045
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2C6 |
Immunogen | Full length human recombit protein of CAMK1D produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: calcium/calmodulin-dependent protein kinase ID; Calcium/calmodulin-dependent protein kinase type 1D; Calcium/calmodulin-dependent protein kinase type 1D CaM kinase ID CaM kinase I delta CaMKI-delta CaM-KI delta CaMKI delta Camk1D CamKI-like protein kinase CKLiK CamKI-like protein kinase CAMK1D; CaM kinase I delta; CaM kinase ID; caM-KI delta; caMKI delta; CaMKI-like protein kinase; CKLiK
Gene Aliases: CaM-K1; CAMK1D; CAMKID; CKLiK
UniProt ID: (Human) Q8IU85
Entrez Gene ID: (Human) 57118
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.