Invitrogen
Manufacturer Code:PA521957
Catalog # PIPA521957
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 385 of CaMK1D (Uniprot IDNo.Q8IU85) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calcium/calmodulin-dependent protein kinase 1D; calcium/calmodulin-dependent protein kinase ID; Calcium/calmodulin-dependent protein kinase type 1D; CaM kinase I delta; CaM kinase ID; CaM-KI delta; caMKI delta; CaMKI-like protein kinase; CKLiK; RP11-462F15.1 CKLiK CaM-K1 CaMKID CaM kinase ID CamKI-like protein kinase caM kinase I delta caM-KI delta caMKI delta calcium/calmodulin-dependent protein kinase type 1D
Gene Aliases: A630059D12Rik; CaM-K1; CAMK1D; CAMKID; CaMKIdelta; CKLiK; E030025C11Rik; mCKLiK
UniProt ID: (Human) Q8IU85, (Mouse) Q8BW96
Entrez Gene ID: (Human) 57118, (Mouse) 227541
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.