Invitrogen
Manufacturer Code:PA141243
Catalog # 01-675-597
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues 10-50 of human CaMK 1D-beta. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calcium/calmodulin-dependent protein kinase ID; Calcium/calmodulin-dependent protein kinase type 1D; CaM kinase I delta; CaM kinase ID; caM-KI delta; caMKI delta; CAMKI MGC120317 MGC120318; CaMKI-like protein kinase; CKLiK
Gene Aliases: CaM-K1; CAMK1D; CAMKID; CKLiK
UniProt ID: (Human) Q8IU85
Entrez Gene ID: (Human) 57118
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.