Abnova
Manufacturer Code:H00000805M01
Catalog # 89-016-409
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3F4-G5 |
Immunogen | CALM2 (AAH03354 1 a.a. ~ 149 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant CALM2.
For Research Use Only
Protein Aliases: calmodulin 2 (phosphorylase kinase, delta); Calmodulin-1; Calmodulin-2; Calmodulin-3; CAMII; LP7057 protein; PHKD; PHKD2; phosphorylase kinase delta; phosphorylase kinase subunit delta; prepro-calmodulin 2
Gene Aliases: CALM; CALM1; CALM2; CALM3; CALML2; CAM; CAM1; CAM2; CAM3; CAMB; CAMC; CAMII; CAMIII; LQT15; PHKD; PHKD2
UniProt ID: (Human) P0DP23
Entrez Gene ID: (Human) 805
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.