Invitrogen
Manufacturer Code:PA5115756
Catalog # PIPA5115756
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PICALM corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CALM; Calm Clathrin assembly lymphoid myeloid leukemia clathrin assembly lymphoid myeloid leukemia protein clathrin-assembly lymphoid leukemia protein clathrin-assembly lymphoid myeloid leukemia protein CLTH fit1 fit-1 LAP mKIAA4114 PAP180 phosphatidylinositol binding clathrin assembly protein phosphatidylinositol-binding clathrin assembly protein Picalm rCALM; Clathrin assembly lymphoid myeloid leukemia; Clathrin assembly lymphoid myeloid leukemia protein; clathrin-assembly lymphoid leukemia protein; clathrin-assembly lymphoid myeloid leukemia protein; Phosphatidylinositol-binding clathrin assembly protein; rCALM
Gene Aliases: CALM; CLTH; fit-1; Fit1; LAP; mKIAA4114; PAP180; PICALM
UniProt ID: (Human) Q13492, (Mouse) Q7M6Y3, (Rat) O55012
Entrez Gene ID: (Human) 8301, (Mouse) 233489, (Rat) 89816
Molecular Function:
membrane traffic protein
vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.