Invitrogen
Manufacturer Code:PA520895
Catalog # PIPA520895
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 17 amino acid peptide near the amino terminus of human CALHM1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium homeostasis modulator protein 1; FAM26C calcium homeostasis modulator protein 1 family with sequence similarity 26 member C; family with sequence similarity 26, member C; Protein FAM26C
Gene Aliases: CALHM1; EG546729; FAM26C; RGD1564891
UniProt ID: (Human) Q8IU99, (Mouse) D3Z291
Entrez Gene ID: (Human) 255022, (Rat) 499367, (Mouse) 546729
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.