Novus Biologicals
Manufacturer Code:NBP292818100UL
Catalog # NB027502
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 202-346 of human CALHM1 (NP_001001412.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calcium homeostasis modulator 1 FAM26Ccalcium homeostasis modulator protein 1 family with sequence similarity 26 member C MGC39514 MGC39617 Protein FAM26C; Calcium homeostasis modulator protein 1; family with sequence similarity 26, member C; Protein FAM26C
Gene Aliases: CALHM1; FAM26C
UniProt ID: (Human) Q8IU99
Entrez Gene ID: (Human) 255022
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.