Proteintech
Manufacturer Code:220421AP150UL
Catalog # 50-173-3699
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CALHM1 Fusion Protein Ag14938 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium homeostasis modulator protein 1; CALHM1 FAM26C Protein FAM26C; family with sequence similarity 26, member C; Protein FAM26C
Gene Aliases: CALHM1; EG546729; FAM26C; RGD1564891
UniProt ID: (Human) Q8IU99, (Mouse) D3Z291
Entrez Gene ID: (Human) 255022, (Rat) 499367, (Mouse) 546729
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.