Invitrogen
Manufacturer Code:PA576414
Catalog # PIPA576414
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human CALCRL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcitonin gene-related peptide type 1 receptor; Calcitonin gene-related peptide type 1 receptor CGRP type 1 receptor Calcitonin receptor-like receptor CALCRL CGRPR; Calcitonin receptor-like receptor; CGRP type 1 receptor
Gene Aliases: AV071593; CALCRL; CGRPR; CRLR
UniProt ID: (Human) Q16602, (Mouse) Q9R1W5
Entrez Gene ID: (Human) 10203, (Mouse) 54598
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.