Invitrogen
Manufacturer Code:PA552511
Catalog # PIPA552511
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CALCRL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcitonin gene-related peptide type 1 receptor; Calcitonin receptor-like receptor; CALCRL CGRP type 1 receptor Cgrp1 receptor CRLR Calcitonin receptor-like CGRP receptor CGRPR; CGRP type 1 receptor
Gene Aliases: CALCRL; CGRPR; CRLR
UniProt ID: (Human) Q16602
Entrez Gene ID: (Human) 10203
Molecular Function:
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.