Abnova
Manufacturer Code:H00000799M01
Catalog # 89-016-408
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2F7 |
Immunogen | CALCR (NP_001733.1 394 a.a. ~ 474 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant CALCR.
For Research Use Only
Protein Aliases: Calcitonin receptor; CRT; CT-R; CTR; CTR1
Gene Aliases: CALCR; CRT; CT-R; CTR; CTR1
UniProt ID: (Human) P30988
Entrez Gene ID: (Human) 799
Molecular Function:
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.