Invitrogen
Manufacturer Code:PA558337
Catalog # PIPA558337
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CALCOCO1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium-binding and coiled-coil domain-containing protein 1; Calphoglin; Cocoa calphoglin calcium-binding and coiled-coil domain-containing protein 1 coiled-coil coactivator protein coiled-coil leucine zipper coactivator 1 inorganic pyrophosphatase activator sarcoma antigen NY-SAR-3; Coiled-coil coactivator protein; coiled-coil leucine zipper coactivator 1; inorganic pyrophosphatase activator; Sarcoma antigen NY-SAR-3
Gene Aliases: CALCOCO1; calphoglin; Cocoa; KIAA1536; PP13275; UNQ2436/PRO4996
UniProt ID: (Human) Q9P1Z2
Entrez Gene ID: (Human) 57658
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.