Invitrogen
Manufacturer Code:PA544788
Catalog # PIPA544788
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human CALCOCO1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium binding and Coiled-coil domain 1 Cocoa KIAA1536 PP13275 calphoglin; Calcium-binding and coiled-coil domain-containing protein 1; Calphoglin; Coiled-coil coactivator protein; coiled-coil leucine zipper coactivator 1; coiled-coil transcriptional coactivator; granule cell antiserum positive 11; GRIP1-interacting protein; inorganic pyrophosphatase activator; Sarcoma antigen NY-SAR-3
Gene Aliases: 1810009B06Rik; CALCOCO1; calphoglin; CocoA; Gcap11; KIAA1536; mKIAA1536; PP13275; UNQ2436/PRO4996
UniProt ID: (Human) Q9P1Z2, (Bovine) Q2KJ21, (Rat) Q66HR5, (Mouse) Q8CGU1
Entrez Gene ID: (Horse) 100064155, (Human) 57658, (Dog) 486505, (Bovine) 538675, (Rabbit) 100337884, (Rat) 246047, (Guinea pig) 100729752, (Mouse) 67488
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.