Abnova
Manufacturer Code:PAB27548
Catalog # 89-129-058
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of human CALB1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of CALB1.
For Research Use Only
Protein Aliases: CALB; Calbindin; calbindin 1, (28kD); calbindin 1, 28kDa; Calbindin D28; D-28K; RTVL-H protein; Vitamin D-dependent calcium-binding protein, avian-type
Gene Aliases: CAB27; CALB; CALB1; D-28K
UniProt ID: (Human) P05937
Entrez Gene ID: (Human) 793
Molecular Function:
calcium-binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.