Invitrogen
Manufacturer Code:PA595979
Catalog # PIPA595979
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 45-344 of human CADM1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CADM1; Cell adhesion molecule 1; cell adhesion molecule 1 transcript variant 8abc; IgSF4; Immunoglobulin superfamily member 4; immunoglobulin superfamily member 4A; immunoglobulin superfamily, member 4; immunoglobulin superfamily, member 4D variant 1; immunoglobulin superfamily, member 4D variant 2; immunosuperfamily protein Bl2; NECL-2; nectin-like 2; Nectin-like protein 2; SgIgSF; Spermatogenic immunoglobulin superfamily; Synaptic cell adhesion molecule; SynCAM; truncated CADM1 protein; TSLC-1; TSLC1/Nectin-like 2/IGSF4; Tumor suppressor in lung cancer 1
Gene Aliases: 2900073G06Rik; 3100001I08Rik; AI987920; BL2; CADM1; IGSF4; IGSF4A; Necl-2; NECL2; RA175; RA175A; RA175B; RA175C; RA175N; SgIGSF; ST17; sTSLC-1; SYNCAM; SynCam1; TSLC1
UniProt ID: (Human) Q9BY67, (Mouse) Q8R5M8
Entrez Gene ID: (Human) 23705, (Mouse) 54725
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.