Invitrogen
Manufacturer Code:PA569508
Catalog # PIPA569508
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of fruit fly CAD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aspartate carbamoyltransferase; CAD protein; CAD protein CAD trifunctional protein multifunctional protein CAD; CAD trifunctional protein; cad-PA; cad-PB; cad-PC; carbamoylphosphate synthetase 2/aspartate transcarbamylase/dihydroorotase; carbamyl phosphatate synthetase 2; CAUDAL; CG1759-PA; CG1759-PB; CG1759-PC; Dihydroorotase; Glutamine-dependent carbamoyl-phosphate synthase; Homeotic protein caudal; LD29596p; multifunctional protein CAD
Gene Aliases: 2410008J01Rik; 38E.19; anon-WO2004063362.83; AU018859; CAD; cb456; cd; CDG1Z; CG1759; Cpad; Dmel\CG1759; Dmel_CG1759; S67; si:dkey-221h15.3; wu:fc30c12; wu:fc33d01; wu:fc67g02
UniProt ID: (Human) P27708, (Mouse) B2RQC6, (Fruit fly) P09085
Entrez Gene ID: (Horse) 100055366, (Rabbit) 100352236, (Bovine) 504261, (Human) 790, (Dog) 483009, (Mouse) 69719, (Guinea pig) 100727133, (Rat) 24240, (Zebrafish) 266992, (Fruit fly) 35341
Molecular Function: ligase transferase DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.