Abnova
Manufacturer Code:PAB17323
Catalog # 89-118-980
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal of human CAD. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of CAD.
For Research Use Only
Protein Aliases: Aspartate carbamoyltransferase; CAD protein; CAD trifunctional protein; Dihydroorotase; Glutamine-dependent carbamoyl-phosphate synthase; multifunctional protein CAD
Gene Aliases: CAD; CDG1Z
UniProt ID: (Human) P27708
Entrez Gene ID: (Human) 790
Molecular Function:
ligase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.