Novus Biologicals
Manufacturer Code:NBP147607IR
Catalog # NBP147607IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S8b-1 |
Immunogen | Synthetic peptide amino acids 189-205 of rat CavBeta2 |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: CAB2; CAB2 CACNLB2 Calcium channel voltage-dependent subunit beta 2 calcium channel voltage-dependent beta 2 subunit CAVB2 FLJ23743 Lambert-Eaton myasthenic syndrome antigen B myasthenic (Lambert-Eaton) syndrome antigen B MYSB voltage-dependent L-type calcium channel subunit beta-2; Calcium channel voltage-dependent subunit beta 2; calcium channel, voltage-dependent, beta 2 subunit; Lambert-Eaton myasthenic syndrome antigen B; myasthenic (Lambert-Eaton) syndrome antigen B; MYSB; Voltage-dependent L-type calcium channel subunit beta-2
Gene Aliases: CACNB2; CACNLB2; CAVB2; MYSB
UniProt ID: (Human) Q5VWV6
Entrez Gene ID: (Human) 783
Molecular Function:
calcium channel
ion channel
transporter
voltage-gated calcium channel
voltage-gated ion channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.