Invitrogen
Manufacturer Code:PA577344
Catalog # PIPA577344
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)HEHVDRYVPHREHNHRE corresponding to amino acid residues 571-587 of rat Cav beta2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAB2; Calcium channel voltage-dependent subunit beta 2; Voltage-dependent L-type calcium channel subunit beta-2; voltage-gated calcium channel beta2 subunit transcript variant X13
Gene Aliases: AW060387; CAB2; Cacnb2; Cacnlb2; Cavbeta2; Cchb2
UniProt ID: (Mouse) Q8CC27, (Rat) Q8VGC3
Entrez Gene ID: (Mouse) 12296, (Rat) 116600
Molecular Function:
calcium channel
ion channel
transporter
voltage-gated calcium channel
voltage-gated ion channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.