Invitrogen
Manufacturer Code:PA576573
Catalog # PIPA576573
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human CACNA2D2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha 2 delta calcium channel subunit; CACNA2D; calcium channel, voltage dependent, alpha2/delta subunit 2; ducky; Protein ducky; Voltage-dependent calcium channel subunit alpha-2-2; Voltage-dependent calcium channel subunit alpha-2/delta-2; Voltage-dependent calcium channel subunit delta-2; Voltage-gated calcium channel subunit alpha-2/delta-2
Gene Aliases: a2d2; Cacna2d; Cacna2d2; du; Kiaa0558; mKIAA0558; td; torpid
UniProt ID: (Mouse) Q6PHS9, (Rat) Q8CFG6
Entrez Gene ID: (Mouse) 56808, (Rat) 300992
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.