Invitrogen
Manufacturer Code:PA535132
Catalog # PIPA535132
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 643-671 amino acids from the central region of human CACNA2D2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha 2 delta calcium channel subunit; CACNA2D2 KIAA0558 Voltage-dependent calcium channel subunit alpha-2/delta-2 Voltage-gated calcium channel subunit alpha-2/delta-2 Voltage-dependent calcium channel subunit alpha-2-2 Voltage-dependent calcium channel subunit delta-2; calcium channel, voltage-dependent, alpha 2/delta subunit 2; ducky; gene 26; Protein ducky; Voltage-dependent calcium channel subunit alpha-2-2; Voltage-dependent calcium channel subunit alpha-2/delta-2; Voltage-dependent calcium channel subunit delta-2; Voltage-gated calcium channel subunit alpha-2/delta-2
Gene Aliases: a2d2; CACNA2D; CACNA2D2; du; KIAA0558; mKIAA0558; td; torpid
UniProt ID: (Human) Q9NY47, (Mouse) Q6PHS9
Entrez Gene ID: (Human) 9254, (Mouse) 56808
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.