Novus Biologicals
Manufacturer Code:NB1202864
Catalog # NB1202864
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 20A |
Immunogen | Purified rabbit dihydropyridine receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CACNA2 CACNL2Adihydropyridine-sensitive L-type calcium channel alpha-2/delta subunit calcium channel L type alpha 2 polypeptide calcium channel voltage-dependent alpha 2/delta subunit 1 CCHL2A MHS3 voltage-dependent calcium channel subunit alpha-2/delta-1 Voltage-gated calcium channel subunit alpha-2/delta-1; calcium channel, L type, alpha 2 polypeptide; calcium channel, voltage-dependent, alpha 2/delta subunit 1; dihydropyridine-sensitive L-type, calcium channel alpha-2/delta subunit; Voltage-dependent calcium channel subunit alpha-2-1; Voltage-dependent calcium channel subunit alpha-2/delta-1; Voltage-dependent calcium channel subunit delta-1; Voltage-gated calcium channel subunit alpha-2/delta-1
Gene Aliases: CACNA2; CACNA2D1; CACNL2A; CCHL2A; LINC01112; lncRNA-N3; MHS3
UniProt ID: (Human) P54289
Entrez Gene ID: (Human) 781
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.