Proteintech
Manufacturer Code:274531AP150UL
Catalog # 50-173-5671
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CACNA2D1 Fusion Protein Ag26190 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CACNA2 CACNA2D1 CACNL2A CCHL2A MHS3; Calcium channel subunit alpha 2 delta (dihydropyridine - sensitive L-type); calcium channel, L type, alpha 2 polypeptide; calcium channel, voltage-dependent, alpha 2/delta subunit 1; dihydropyridine-sensitive calcium channel alpha 2; dihydropyridine-sensitive L-type, calcium channel alpha-2/delta subunit; Voltage-dependent calcium channel subunit alpha-2-1; Voltage-dependent calcium channel subunit alpha-2/delta-1; Voltage-dependent calcium channel subunit delta-1; Voltage-gated calcium channel subunit alpha-2/delta-1
Gene Aliases: Ca(v)alpha2delta1; CACNA2; CACNA2D1; CACNL2A; CCHL2A; CCHLA2; DHSCCA; LINC01112; lncRNA-N3; MHS3
UniProt ID: (Human) P54289, (Mouse) O08532
Entrez Gene ID: (Pig) 397377, (Human) 781, (Rat) 25399, (Mouse) 12293
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.