Abnova
Manufacturer Code:MAB6650
Catalog # 89-112-976
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S55-10 |
Immunogen | Recombinant fusion protein corresponding to amino acids 1019-1293 (II-III loop) of human CACNA1H. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant CACNA1H.
For Research Use Only
Protein Aliases: CACNA1HB; calcium channel, voltage-dependent, T type, alpha 1H subunit; calcium channel, voltage-dependent, T type, alpha 1Hb subunit; Cav3.2; EIG6; FLJ90484; Low-voltage-activated calcium channel alpha1 3.2 subunit; low-voltage-activated calcium channel alpha13.2 subunit; voltage dependent t-type calcium channel alpha-1H subunit; Voltage-dependent T-type calcium channel subunit alpha-1H; voltage-gated calcium channel alpha subunit Cav3.2; voltage-gated calcium channel alpha subunit CavT.2; Voltage-gated calcium channel subunit alpha Cav3.2
Gene Aliases: CACNA1H; CACNA1HB; Cav3.2; ECA6; EIG6
UniProt ID: (Human) O95180
Entrez Gene ID: (Human) 8912
Molecular Function:
calcium channel
ion channel
sodium channel
transporter
voltage-gated calcium channel
voltage-gated ion channel
voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.