Abnova
Manufacturer Code:PAB20277
Catalog # 89-121-781
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human CACNA1G. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant CACNA1G.
For Research Use Only
Protein Aliases: Ca(V)T.1; calcium channel, voltage-dependent, T type, alpha 1G subunit; Cav3.1; Cav3.1c; MGC117234; NBR13; voltage-dependent calcium channel alpha 1G subunit, isoform 11; voltage-dependent T-type calcium channel alpha 1G subunit; Voltage-dependent T-type calcium channel subunit alpha-1G; Voltage-gated calcium channel subunit alpha Cav3.1
Gene Aliases: Ca(V)T.1; CACNA1G; Cav3.1; KIAA1123; NBR13; SCA42
UniProt ID: (Human) Q19R12
Entrez Gene ID: (Human) 8913
Molecular Function: calcium channel ion channel sodium channel transporter voltage-gated calcium channel voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.