Novus Biologicals
Manufacturer Code:NBP238049
Catalog # NBP238049
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: KFHTTCFEEGTDDIQGESPAPCGTEEPARTCPNGTKCQPYWEGPNNGI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APCA BI brain calcium channel 1 Brain calcium channel I CACH4 CACN3 CACNL1A4CAV2.1 calcium channel L type alpha-1 polypeptide Calcium channel L type alpha-1 polypeptide isoform 4 calcium channel voltage-dependent P/Q type alpha 1A subunit Cav2.1 EA2 FHM HPCA MHP MHP1 SCA6 voltage-dependent P/Q-type calcium channel subunit alpha-1A Voltage-gated calcium channel subunit alpha Cav2.1; BI; brain calcium channel 1; Brain calcium channel I; calcium channel, L type, alpha-1 polypeptide; Calcium channel, L type, alpha-1 polypeptide isoform 4; calcium channel, voltage-dependent, P/Q type, alpha 1A subunit; fetal brain Ca2+ voltage-gated channel alpha1A pore-forming subunit; Voltage-dependent P/Q-type calcium channel subunit alpha-1A; Voltage-gated calcium channel subunit alpha Cav2.1
Gene Aliases: APCA; BI; CACH4; CACN3; CACNA1A; CACNL1A4; CAV2.1; EA2; FHM; HPCA; MHP; MHP1; SCA6
UniProt ID: (Human) O00555
Entrez Gene ID: (Human) 773
Molecular Function:
calcium channel
ion channel
sodium channel
transporter
voltage-gated calcium channel
voltage-gated ion channel
voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.